Assay Details
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289462.1 | NP_001276391.1 | 3 - 4 | 650 | 87 | |
NM_001289463.1 | NP_001276392.1 | 3 - 4 | 434 | 87 | ||
NM_008604.4 | NP_032630.2 | 3 - 4 | 284 | 87 | ||
GenBank mRNA | AK031446.1 | - | 3 - 4 | 285 | 87 | |
AK033824.1 | - | 3 - 4 | 345 | 87 | ||
AK040730.1 | - | 3 - 4 | 321 | 87 | ||
AK052088.1 | - | 2 - 3 | 171 | 87 | ||
AK076908.1 | - | 3 - 4 | 498 | 87 | ||
AK080202.1 | - | 3 - 4 | 312 | 87 | ||
BC034092.1 | - | 3 - 4 | 306 | 4483558 | 87 | |
BC066840.1 | - | 2 - 3 | 193 | 87 | ||
M81591.1 | - | 2 - 3 | 196 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
