Assay Details
Gene Symbol: Cyp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009993.3 | NP_034123.1 | 4 - 5 | 1093 | 76 | |
GenBank mRNA | BC018298.1 | - | 4 - 5 | 1073 | 4196676 | 76 |
BC054827.1 | - | 4 - 5 | 1086 | 5054733 | 76 | |
K02589.1 | - | 4 - 5 | 1094 | 76 | ||
X00479.1 | - | 4 - 5 | 1094 | 76 | ||
X04283.1 | - | 3 - 4 | 1057 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
