Assay Details
Gene Symbol: Rfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011258.2 | NP_035388.2 | 7 - 8 | 833 | 120 | |
GenBank mRNA | AK136937.1 | - | 7 - 8 | 812 | 120 | |
AK146164.1 | - | 7 - 8 | 828 | 120 | ||
AK146314.1 | - | 7 - 8 | 828 | 120 | ||
BC085173.1 | - | 7 - 8 | 810 | 30549691 | 120 | |
BC085174.1 | - | 7 - 8 | 802 | 120 | ||
U01222.1 | - | 7 - 8 | 801 | 120 | ||
U07157.1 | - | 7 - 8 | 799 | 120 | ||
U15037.1 | - | 7 - 8 | 834 | 120 | ||
U36441.1 | - | 7 - 8 | 810 | 120 | ||
X72711.1 | - | 7 - 8 | 799 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
