Assay Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 4 - 5 | 759 | 90 | |
NM_001110351.1 | NP_001103821.1 | 5 - 6 | 636 | 90 | ||
NM_011378.2 | NP_035508.2 | 5 - 6 | 867 | 90 | ||
GenBank mRNA | AK032209.1 | - | 4 - 5 | 627 | 90 | |
AK220292.1 | - | 4 - 5 | 736 | 90 | ||
BC052716.1 | - | 4 - 5 | 646 | 6837144 | 90 | |
BC053385.1 | - | 4 - 5 | 90 | |||
L38620.2 | - | 3 - 4 | 526 | 90 | ||
L38621.2 | - | 3 - 4 | 526 | 90 | ||
U22394.1 | - | 5 - 6 | 875 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
