Assay Details
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113559.2 | NP_001107031.2 | 10 - 11 | 1541 | 61 | |
NM_001243163.1 | NP_001230092.1 | 10 - 11 | 1433 | 61 | ||
NM_011444.3 | NP_035574.2 | 11 - 12 | 1685 | 61 | ||
GenBank mRNA | AB006330.1 | - | 11 - 12 | 1597 | 61 | |
AJ010604.1 | - | 10 - 11 | 1423 | 61 | ||
AK015212.1 | - | 4 - 5 | 494 | 61 | ||
AK029047.1 | - | 13 - 14 | 1683 | 61 | ||
AK160154.1 | - | 4 - 5 | 499 | 61 | ||
BC110478.2 | - | 10 - 11 | 1425 | 40047865 | 61 | |
KF793916.1 | - | 11 - 12 | 1386 | 61 | ||
X65657.1 | - | 4 - 5 | 470 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
