Assay Details
Gene Symbol: Cngb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013927.2 | NP_038955.1 | 2 - 3 | 271 | 104 | |
GenBank mRNA | AJ243572.1 | - | 2 - 3 | 271 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
