Assay Details
Gene Symbol: Skap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018773.2 | NP_061243.1 | 2 - 3 | 321 | 98 | |
GenBank mRNA | AB014485.1 | - | 2 - 3 | 238 | 98 | |
AF051324.1 | - | 2 - 3 | 236 | 98 | ||
AK076000.1 | - | 2 - 3 | 266 | 98 | ||
AK144289.1 | - | 2 - 3 | 258 | 98 | ||
AK172297.1 | - | 2 - 3 | 305 | 98 | ||
BC003711.1 | - | 2 - 3 | 321 | 3599914 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
