Assay Details
Gene Symbol: Tnfaip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009398.2 | NP_033424.1 | 2 - 3 | 292 | 67 | |
GenBank mRNA | AK139985.1 | - | 2 - 3 | 302 | 67 | |
BC021155.1 | - | 2 - 3 | 271 | 4208738 | 67 | |
U83903.1 | - | 2 - 3 | 292 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
