Assay Details
Gene Symbol: Traf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009423.4 | NP_033449.2 | 1 - 2 | 253 | 128 | |
GenBank mRNA | AF233449.1 | - | 1 - 2 | 152 | 128 | |
AK035056.1 | - | 1 - 2 | 231 | 128 | ||
AK040877.1 | - | 231 | 128 | |||
AK046015.1 | - | 1 - 2 | 293 | 128 | ||
AK053900.1 | - | 1 - 2 | 250 | 128 | ||
BC020455.1 | - | 1 - 2 | 238 | 128 | ||
X92346.1 | - | 1 - 2 | 273 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
