Assay Details
Gene Symbol: Uba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136085.2 | NP_001129557.1 | 10 - 11 | 1285 | 124 | |
NM_001276316.1 | NP_001263245.1 | 11 - 12 | 1377 | 124 | ||
NM_001276317.1 | NP_001263246.1 | 10 - 11 | 1280 | 124 | ||
NM_009457.4 | NP_033483.2 | 10 - 11 | 1280 | 124 | ||
GenBank mRNA | AK088057.1 | - | 10 - 11 | 1162 | 124 | |
AK088528.1 | - | 10 - 11 | 1162 | 124 | ||
AK143416.1 | - | 10 - 11 | 1186 | 124 | ||
AK171667.1 | - | 10 - 11 | 1251 | 124 | ||
AK185680.1 | - | 1 - 2 | 120 | 124 | ||
AK195016.1 | - | 1 - 2 | 108 | 124 | ||
AK206443.1 | - | 1 - 2 | 52 | 124 | ||
AK210744.1 | - | 1 - 2 | 109 | 124 | ||
BC058630.1 | - | 10 - 11 | 1226 | 6402368 | 124 | |
BC138200.1 | - | 10 - 11 | 1143 | 8861219 | 124 | |
BC145984.1 | - | 10 - 11 | 1144 | 40131287 | 124 | |
D10576.1 | - | 10 - 11 | 1146 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
