Assay Details
Gene Symbol: Dmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010059.3 | NP_034189.1 | 7 - 8 | 600 | 93 | |
GenBank mRNA | BC116767.1 | - | 7 - 8 | 485 | 40126086 | 93 |
BC119081.1 | - | 7 - 8 | 485 | 8733830 | 93 | |
D58419.1 | - | 7 - 8 | 585 | 93 | ||
D64107.1 | - | 7 - 8 | 574 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
