Assay Details
Gene Symbol: Ica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010492.3 | NP_034622.3 | 11 - 12 | 1493 | 95 | |
GenBank mRNA | AB303567.1 | - | 10 - 11 | 1004 | 95 | |
BC002030.1 | - | 11 - 12 | 1127 | 3487814 | 95 | |
DV658174.1 | - | 5 - 6 | 403 | 95 | ||
U26459.1 | - | 11 - 12 | 1116 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
