Assay Details
Gene Symbol: Traf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011633.2 | NP_035763.2 | 562 | 99 | ||
GenBank mRNA | AK141097.1 | - | 2 - 3 | 523 | 99 | |
AK144694.1 | - | 513 | 99 | |||
BC012702.1 | - | 516 | 4218625 | 99 | ||
D78141.1 | - | 539 | 99 | |||
D83528.1 | - | 404 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
