Assay Details
Gene Symbol: Traf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011633.2 | NP_035763.2 | 889 | 77 | ||
GenBank mRNA | AK141097.1 | - | 5 - 6 | 850 | 77 | |
AK144694.1 | - | 840 | 77 | |||
BC012702.1 | - | 843 | 4218625 | 77 | ||
D78141.1 | - | 866 | 77 | |||
D83528.1 | - | 731 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
