Assay Details
Gene Symbol: Traf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011633.2 | NP_035763.2 | 1279 | 126 | ||
GenBank mRNA | AK144694.1 | - | 1230 | 126 | ||
BC012702.1 | - | 1233 | 4218625 | 126 | ||
D78141.1 | - | 1256 | 126 | |||
D83528.1 | - | 1121 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
