Assay Details
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013529.3 | NP_038557.1 | 6 - 7 | 617 | 75 | |
GenBank mRNA | AB016780.1 | - | 6 - 7 | 583 | 75 | |
AK132821.1 | - | 5 - 6 | 463 | 75 | ||
BC031928.1 | - | 6 - 7 | 585 | 4167189 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
