Assay Details
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013529.3 | NP_038557.1 | 9 - 10 | 881 | 62 | |
GenBank mRNA | AB016780.1 | - | 9 - 10 | 847 | 62 | |
AK132821.1 | - | 8 - 9 | 727 | 62 | ||
BC031928.1 | - | 9 - 10 | 849 | 4167189 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
