Assay Details
Gene Symbol: Txnrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013711.3 | NP_038739.2 | 6 - 7 | 565 | 57 | |
GenBank mRNA | AB027566.1 | - | 6 - 7 | 565 | 57 | |
AF136399.1 | - | 6 - 7 | 568 | 57 | ||
AF171053.1 | - | 6 - 7 | 531 | 57 | ||
AF412308.1 | - | 6 - 7 | 567 | 57 | ||
AK075828.1 | - | 6 - 7 | 543 | 57 | ||
BC013688.1 | - | 6 - 7 | 545 | 3584154 | 57 | |
BC029344.1 | - | 6 - 7 | 545 | 57 | ||
BC052157.1 | - | 6 - 7 | 544 | 6506286 | 57 | |
BC052758.1 | - | 6 - 7 | 544 | 57 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
