Assay Details
Gene Symbol: Tjp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282095.1 | NP_001269024.1 | 5 - 6 | 720 | 84 | |
NM_001282096.1 | NP_001269025.1 | 5 - 6 | 783 | 84 | ||
NM_013769.3 | NP_038797.2 | 5 - 6 | 822 | 84 | ||
GenBank mRNA | AF157006.1 | - | 5 - 6 | 790 | 84 | |
AK029083.1 | - | 5 - 6 | 769 | 84 | ||
AK134100.1 | - | 5 - 6 | 823 | 84 | ||
AK167587.1 | - | 5 - 6 | 721 | 84 | ||
BC012518.1 | - | 5 - 6 | 787 | 3600848 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
