Assay Details
Gene Symbol: Abcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013806.2 | NP_038834.2 | 19 - 20 | 2708 | 62 | |
GenBank mRNA | AF227274.1 | - | 19 - 20 | 2612 | 62 | |
AF282772.1 | - | 19 - 20 | 2612 | 62 | ||
AF282773.1 | - | 19 - 20 | 2612 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
