Assay Details
Gene Symbol: Itgax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021334.2 | NP_067309.1 | 19 - 20 | 2422 | 84 | |
GenBank mRNA | AF211864.1 | - | 19 - 20 | 2422 | 84 | |
AK131133.1 | - | 19 - 20 | 2428 | 84 | ||
AK155816.1 | - | 19 - 20 | 2444 | 84 | ||
AK170432.1 | - | 19 - 20 | 2440 | 84 | ||
BC042840.1 | - | 19 - 20 | 2421 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
