Assay Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162500.1 | NP_001155972.1 | 8 - 9 | 758 | 71 | |
NM_022321.4 | NP_071716.3 | 8 - 9 | 713 | 71 | ||
GenBank mRNA | AF312712.1 | - | 8 - 9 | 671 | 71 | |
AK028329.1 | - | 8 - 9 | 714 | 71 | ||
AK037365.1 | - | 8 - 9 | 686 | 71 | ||
AK040914.1 | - | 8 - 9 | 644 | 71 | ||
AK079881.1 | - | 8 - 9 | 639 | 71 | ||
AK088419.1 | - | 8 - 9 | 663 | 71 | ||
BC011200.1 | - | 8 - 9 | 667 | 4190510 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
