Assay Details
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025768.2 | NP_080044.2 | 6 - 7 | 856 | 64 | |
GenBank mRNA | AK007905.1 | - | 6 - 7 | 861 | 64 | |
AK017197.1 | - | 6 - 7 | 820 | 64 | ||
AK017251.1 | - | 7 - 8 | 818 | 64 | ||
AK082863.1 | - | 6 - 7 | 850 | 64 | ||
AK149322.1 | - | 853 | 64 | |||
BC029096.1 | - | 6 - 7 | 862 | 3500563 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
