Assay Details
Gene Symbol: Lztr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025808.3 | NP_080084.2 | 15 - 16 | 1834 | 81 | |
GenBank mRNA | AK004561.2 | - | 15 - 16 | 1834 | 81 | |
AK005037.1 | - | 15 - 16 | 1833 | 81 | ||
AK083411.1 | - | 15 - 16 | 1838 | 81 | ||
AK088938.1 | - | 6 - 7 | 766 | 81 | ||
AK132817.1 | - | 15 - 16 | 1833 | 81 | ||
BC034400.1 | - | 15 - 16 | 1828 | 2651456 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
