Assay Details
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025951.3 | NP_080227.2 | 5 - 6 | 1019 | 122 | |
NM_028744.3 | NP_083020.2 | 5 - 6 | 950 | 122 | ||
GenBank mRNA | AF411321.1 | - | 5 - 6 | 874 | 122 | |
AK011751.1 | - | 5 - 6 | 982 | 122 | ||
AK016754.1 | - | 5 - 6 | 913 | 122 | ||
AK035523.1 | - | 5 - 6 | 1004 | 122 | ||
AK170730.1 | - | 5 - 6 | 1003 | 122 | ||
AY148879.1 | - | 913 | 122 | |||
BC062144.1 | - | 5 - 6 | 926 | 6335661 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
