Assay Details
Gene Symbol: Rpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164223.1 | NP_001157695.1 | 7 - 8 | 669 | 81 | |
NM_026653.2 | NP_080929.1 | 6 - 7 | 606 | 81 | ||
GenBank mRNA | AK035084.1 | - | 6 - 7 | 534 | 81 | |
AK148035.1 | - | 6 - 7 | 547 | 81 | ||
AK150785.1 | - | 6 - 7 | 503 | 81 | ||
AK151590.1 | - | 6 - 7 | 559 | 81 | ||
AK152296.1 | - | 7 - 8 | 599 | 81 | ||
AK153107.1 | - | 7 - 8 | 599 | 81 | ||
AK153144.1 | - | 6 - 7 | 559 | 81 | ||
AK165316.1 | - | 6 - 7 | 519 | 81 | ||
AK165944.1 | - | 6 - 7 | 537 | 81 | ||
AK167598.1 | - | 6 - 7 | 492 | 81 | ||
AK168400.1 | - | 6 - 7 | 558 | 81 | ||
AK169599.1 | - | 6 - 7 | 530 | 81 | ||
AK187897.1 | - | 2 - 3 | 164 | 81 | ||
AK189517.1 | - | 2 - 3 | 161 | 81 | ||
AK197921.1 | - | 2 - 3 | 161 | 81 | ||
AK206519.1 | - | 1 - 2 | 45 | 81 | ||
BC019119.1 | - | 6 - 7 | 537 | 4487586 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
