Assay Details
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028744.3 | NP_083020.2 | 2 - 3 | 466 | 113 | |
GenBank mRNA | AK016754.1 | - | 2 - 3 | 432 | 113 | |
AY148879.1 | - | 432 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
