Assay Details
Gene Symbol: Pask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080850.2 | NP_543126.2 | 6 - 7 | 1186 | 60 | |
GenBank mRNA | AK028435.1 | - | 6 - 7 | 1188 | 60 | |
AK083528.1 | - | 6 - 7 | 1157 | 60 | ||
AK083590.1 | - | 6 - 7 | 1151 | 60 | ||
AK129063.1 | - | 6 - 7 | 1076 | 60 | ||
AK172031.1 | - | 6 - 7 | 1134 | 60 | ||
BC150821.1 | - | 5 - 6 | 898 | 9087732 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
