Assay Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080853.3 | NP_543129.3 | 6 - 7 | 1558 | 82 | |
GenBank mRNA | AF324864.1 | - | 6 - 7 | 1391 | 82 | |
AK043753.1 | - | 5 - 6 | 1011 | 82 | ||
AK045409.1 | - | 6 - 7 | 1321 | 82 | ||
BC038375.1 | - | 6 - 7 | 937 | 5357143 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
