Assay Details
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008628.2 | NP_032654.1 | 5 - 6 | 983 | 60 | |
GenBank mRNA | AK051960.1 | - | 5 - 6 | 980 | 60 | |
AK157842.1 | - | 5 - 6 | 996 | 60 | ||
AK166179.1 | - | 5 - 6 | 969 | 60 | ||
BC047117.1 | - | 5 - 6 | 991 | 4951173 | 60 | |
BC050897.1 | - | 5 - 6 | 966 | 6401835 | 60 | |
U21011.1 | - | 5 - 6 | 1000 | 60 | ||
X81143.1 | - | 5 - 6 | 983 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
