Assay Details
Gene Symbol: Plxna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008883.2 | NP_032909.2 | 6 - 7 | 1734 | 85 | |
GenBank mRNA | AK220285.1 | - | 4 - 5 | 459 | 85 | |
BC093482.1 | - | 6 - 7 | 1717 | 30945372 | 85 | |
D86950.1 | - | 6 - 7 | 1700 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
