Assay Details
Gene Symbol: Copg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017477.2 | NP_059505.1 | 9 - 10 | 836 | 67 | |
GenBank mRNA | AF187079.1 | - | 9 - 10 | 835 | 67 | |
AK049417.1 | - | 9 - 10 | 787 | 67 | ||
AK077461.1 | - | 9 - 10 | 783 | 67 | ||
AK147895.1 | - | 9 - 10 | 849 | 67 | ||
AK151284.1 | - | 9 - 10 | 841 | 67 | ||
AK151816.1 | - | 9 - 10 | 841 | 67 | ||
AK153142.1 | - | 9 - 10 | 841 | 67 | ||
AK164280.1 | - | 9 - 10 | 806 | 67 | ||
AK169256.1 | - | 9 - 10 | 881 | 67 | ||
AK171950.1 | - | 9 - 10 | 815 | 67 | ||
AK172519.1 | - | 9 - 10 | 852 | 67 | ||
BC024686.1 | - | 9 - 10 | 807 | 3992144 | 67 | |
BC024896.1 | - | 9 - 10 | 804 | 4189311 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
