Assay Details
Gene Symbol: Ercc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133658.1 | NP_598419.1 | 8 - 9 | 1414 | 62 | |
GenBank mRNA | AK151188.1 | - | 8 - 9 | 1419 | 62 | |
AK152347.1 | - | 8 - 9 | 1419 | 62 | ||
AK152988.1 | - | 8 - 9 | 1426 | 62 | ||
AK154745.1 | - | 8 - 9 | 1420 | 62 | ||
AK160187.1 | - | 2 - 3 | 518 | 62 | ||
BC016595.1 | - | 8 - 9 | 1385 | 3153847 | 62 | |
BC022956.1 | - | 7 - 8 | 1518 | 62 | ||
BC026575.1 | - | 8 - 9 | 1396 | 4955015 | 62 | |
S71186.1 | - | 8 - 9 | 1414 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
