Assay Details
Gene Symbol: Ercc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133658.1 | NP_598419.1 | 13 - 14 | 2130 | 82 | |
GenBank mRNA | AK151188.1 | - | 13 - 14 | 2135 | 82 | |
AK152347.1 | - | 13 - 14 | 2135 | 82 | ||
AK152988.1 | - | 13 - 14 | 2142 | 82 | ||
AK160187.1 | - | 7 - 8 | 1234 | 82 | ||
BC016595.1 | - | 13 - 14 | 2101 | 3153847 | 82 | |
BC022956.1 | - | 12 - 13 | 2234 | 82 | ||
BC024446.1 | - | 4 - 5 | 476 | 82 | ||
BC026575.1 | - | 13 - 14 | 2112 | 4955015 | 82 | |
S71186.1 | - | 13 - 14 | 2130 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
