Assay Details
Gene Symbol: Dcaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199009.1 | NP_001185938.1 | 5 - 6 | 744 | 148 | |
NM_133734.3 | NP_598495.1 | 5 - 6 | 649 | 148 | ||
NR_037572.1 | 5 - 6 | 761 | 148 | |||
GenBank mRNA | AK033540.1 | - | 5 - 6 | 647 | 148 | |
AK152395.1 | - | 4 - 5 | 702 | 148 | ||
AK158784.1 | - | 5 - 6 | 729 | 148 | ||
AK170657.1 | - | 5 - 6 | 647 | 148 | ||
BC008545.1 | - | 4 - 5 | 542 | 3497641 | 148 | |
BC021305.1 | - | 3 - 4 | 2241 | 148 | ||
BC037001.1 | - | 4 - 5 | 673 | 5051353 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
