Assay Details
Gene Symbol: Ssbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133772.2 | NP_598533.2 | 11 - 12 | 851 | 83 | |
GenBank mRNA | AK004835.1 | - | 11 - 12 | 964 | 83 | |
AK154336.1 | - | 11 - 12 | 852 | 83 | ||
AK183440.1 | - | 1 - 2 | 43 | 83 | ||
AK188774.1 | - | 1 - 2 | 43 | 83 | ||
BC002150.1 | - | 11 - 12 | 810 | 3484896 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
