Assay Details
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285872.1 | NP_001272801.1 | 2 - 3 | 245 | 64 | |
NM_001285874.1 | NP_001272803.1 | 1 - 2 | 187 | 64 | ||
NM_001285875.1 | NP_001272804.1 | 2 - 3 | 234 | 64 | ||
NM_133880.3 | NP_598641.2 | 2 - 3 | 234 | 64 | ||
GenBank mRNA | AK144090.1 | - | 2 - 3 | 197 | 64 | |
AK145092.1 | - | 2 - 3 | 185 | 64 | ||
AK156774.1 | - | 1 - 2 | 188 | 64 | ||
BC021890.1 | - | 1 - 2 | 139 | 3598000 | 64 | |
BC025495.1 | - | 2 - 3 | 256 | 5310200 | 64 | |
BC025938.1 | - | 1 - 2 | 80 | 64 | ||
BY233826.1 | - | 1 - 2 | 181 | 64 | ||
BY782149.1 | - | 2 - 3 | 235 | 64 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001285875.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
