Assay Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134253.2 | NP_599014.2 | 7 - 8 | 1028 | 61 | |
GenBank mRNA | BC005659.1 | - | 7 - 8 | 937 | 3588701 | 61 |
BC006585.1 | - | 3 - 4 | 284 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
