Assay Details
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199186.1 | NP_001186115.1 | 5 - 6 | 428 | 60 | |
NM_152808.3 | NP_690021.1 | 5 - 6 | 430 | 60 | ||
GenBank mRNA | AK029457.1 | - | 5 - 6 | 390 | 60 | |
AK041533.1 | - | 5 - 6 | 387 | 60 | ||
AK043924.1 | - | 5 - 6 | 400 | 60 | ||
AK048648.1 | - | 5 - 6 | 415 | 60 | ||
AK088553.1 | - | 5 - 6 | 377 | 60 | ||
AK153992.1 | - | 5 - 6 | 409 | 60 | ||
AK166695.1 | - | 5 - 6 | 431 | 60 | ||
BC023763.1 | - | 5 - 6 | 413 | 60 | ||
BC031535.1 | - | 5 - 6 | 406 | 3991609 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
