Assay Details
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023617.2 | NP_076106.2 | 24 - 25 | 2763 | 149 | |
GenBank mRNA | AF172276.2 | - | 24 - 25 | 2839 | 149 | |
AK004700.1 | - | 13 - 14 | 1559 | 149 | ||
AK030109.1 | - | 20 - 21 | 2303 | 149 | ||
AK149434.1 | - | 24 - 25 | 2724 | 149 | ||
AY347569.1 | - | 24 - 25 | 2640 | 149 | ||
BC049853.1 | - | 24 - 25 | 2715 | 149 | ||
BC138876.1 | - | 24 - 25 | 2742 | 8861898 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
