Assay Details
Gene Symbol: Tnfaip8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027206.2 | NP_081482.1 | 1 - 2 | 46 | 78 | |
GenBank mRNA | AK007540.1 | - | 1 - 2 | 47 | 78 | |
BC094616.1 | - | 1 - 2 | 50 | 573953 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK007540.1,BC094616.1,NM_027206.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342445 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342446 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150678 | BC094616.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150678 | NM_027206.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150678 | AK007540.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150678 | BC055879.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150678 | BC094616.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150678 | NM_027206.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150678 | AK007540.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150678 | BC055879.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78625 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512483 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30331 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158575 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150678 | BC094616.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150678 | NM_027206.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150678 | AK007540.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150678 | BC055879.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150678 | BC094616.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150678 | NM_027206.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150678 | AK007540.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150678 | BC055879.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342446 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342445 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342442 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342449 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342485 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342454 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22342467 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158575 | - | - |
Whole Mouse Genome G4122A | A_51_P150678 | BC094616.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P150678 | NM_027206.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P150678 | AK007540.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P150678 | BC055879.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P150678 | BC094616.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P150678 | NM_027206.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P150678 | AK007540.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P150678 | BC055879.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100862 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158575 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02368730 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02330333 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02368739 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02368740 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02368732 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342445 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22342446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22342382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342487 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02649858 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03125478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342465 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342470 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02409296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02368744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342468 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02649856 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342485 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342454 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342449 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342442 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22342461 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10500102 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500101 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452948_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10500100 | - | - |
Mouse Genome 430 2.0 Array | 1452948_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452948_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69769_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69769_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300019053.mm.1 | NM_027206.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019053.mm.1 | NM_027206.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019053.mm.1 | TC0300002467.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452948_PM_at | - | - |
Mouse Expression Set 430 | 1452948_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452948_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1452948_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 697613 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 7000600 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 7000600 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007000600 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249688 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1249688 | - | - |
mouse-6 v1.1 (Illumina) | scl21847.2.1_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7000600 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl21847.2.1_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7000600 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1249688 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2878979 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249688 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2878979 | - | - |
