Assay Details
Gene Symbol: Tbc1d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144517.4 | NP_653100.1 | 1 - 2 | 234 | 74 | |
GenBank mRNA | AK012836.1 | - | 1 - 2 | 186 | 74 | |
AK076128.1 | - | 1 - 2 | 182 | 74 | ||
AK083084.1 | - | 1 - 2 | 207 | 74 | ||
AK177065.1 | - | 1 - 2 | 119 | 74 | ||
BC020138.1 | - | 1 - 2 | 193 | 3983701 | 74 | |
BY127335.1 | - | 1 - 2 | 234 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
