Assay Details
Gene Symbol: Nae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144931.3 | NP_659180.1 | 18 - 19 | 1520 | 84 | |
GenBank mRNA | AK146202.1 | - | 18 - 19 | 1478 | 84 | |
BC019163.1 | - | 18 - 19 | 1500 | 3711115 | 84 | |
BC021510.1 | - | 18 - 19 | 1501 | 5347277 | 84 | |
BC023680.1 | - | 18 - 19 | 1502 | 5343341 | 84 | |
BC023897.1 | - | 18 - 19 | 1477 | 5321187 | 84 | |
BC027124.1 | - | 18 - 19 | 1464 | 5355631 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
