Assay Details
Gene Symbol: Apeh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146226.2 | NP_666338.2 | 15 - 16 | 1475 | 64 | |
GenBank mRNA | AK149671.1 | - | 15 - 16 | 1479 | 64 | |
AK149713.1 | - | 15 - 16 | 1477 | 64 | ||
AK217552.1 | - | 2 - 3 | 222 | 64 | ||
BC005716.1 | - | 14 - 15 | 1601 | 64 | ||
BC023937.1 | - | 5 - 6 | 444 | 64 | ||
BC025494.1 | - | 15 - 16 | 1457 | 5310043 | 64 | |
BC026594.1 | - | 10 - 11 | 1008 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
