Assay Details
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026214.2 | NP_001021385.1 | 9 - 10 | 908 | 62 | |
NM_001286049.1 | NP_001272978.1 | 8 - 9 | 883 | 62 | ||
NM_001286058.1 | NP_001272987.1 | 7 - 8 | 810 | 62 | ||
NM_007647.3 | NP_031673.2 | 8 - 9 | 835 | 62 | ||
GenBank mRNA | AF006482.1 | - | 8 - 9 | 843 | 62 | |
AF136571.1 | - | 8 - 9 | 869 | 62 | ||
AJ238636.1 | - | 8 - 9 | 871 | 62 | ||
AK002618.1 | - | 8 - 9 | 808 | 62 | ||
AK036641.1 | - | 9 - 10 | 885 | 62 | ||
AK037736.1 | - | 8 - 9 | 804 | 62 | ||
AK045828.1 | - | 8 - 9 | 821 | 62 | ||
AK079267.1 | - | 7 - 8 | 732 | 62 | ||
AK080265.1 | - | 9 - 10 | 895 | 62 | ||
AK081435.1 | - | 9 - 10 | 873 | 62 | ||
AK088455.1 | - | 8 - 9 | 864 | 62 | ||
AK160950.1 | - | 6 - 7 | 729 | 62 | ||
AK163688.1 | - | 8 - 9 | 809 | 62 | ||
BC015247.1 | - | 7 - 8 | 793 | 4242560 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
