Assay Details
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009602.4 | NP_033732.2 | 2 - 3 | 435 | 64 | |
GenBank mRNA | AF089739.1 | - | 1 - 2 | 125 | 64 | |
AF145286.1 | - | 2 - 3 | 374 | 64 | ||
AF299083.1 | - | 2 - 3 | 229 | 64 | ||
AF325349.1 | - | 2 - 3 | 195 | 64 | ||
AK049722.1 | - | 2 - 3 | 408 | 64 | ||
AK051742.1 | - | 2 - 3 | 417 | 64 | ||
AK081254.1 | - | 412 | 64 | |||
AY574267.1 | - | 2 - 3 | 209 | 64 | ||
BC065103.1 | - | 2 - 3 | 388 | 6838257 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
