Assay Details
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010269.3 | NP_034399.1 | 1 - 2 | 205 | 116 | |
NR_104341.1 | 1 - 2 | 205 | 116 | |||
GenBank mRNA | AK004887.1 | - | 1 - 2 | 151 | 116 | |
AK082958.1 | - | 1 - 2 | 167 | 116 | ||
AK146835.1 | - | 1 - 2 | 195 | 116 | ||
AK161122.1 | - | 1 - 2 | 142 | 116 | ||
AK166799.1 | - | 1 - 2 | 120 | 116 | ||
AK166859.1 | - | 1 - 2 | 119 | 116 | ||
BY084929.1 | - | 1 - 2 | 207 | 117 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK004887.1, AK082958.1, AK146835.1, AK166799.1, AK166859.1, NM_010269.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
