Assay Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF402611.1 | - | 10 - 11 | 1666 | 66 | |
AF402612.1 | - | 9 - 10 | 1563 | 66 | ||
AF402613.1 | - | 10 - 11 | 1713 | 66 | ||
AK028302.1 | - | 7 - 8 | 1115 | 66 | ||
AK034504.1 | - | 9 - 10 | 1656 | 66 | ||
AK134961.1 | - | 10 - 11 | 1729 | 66 | ||
AK149626.1 | - | 10 - 11 | 1717 | 66 | ||
BC129902.1 | - | 10 - 11 | 1753 | 40111637 | 66 | |
BC129903.1 | - | 10 - 11 | 1754 | 40111639 | 66 | |
BC145523.1 | - | 10 - 11 | 1807 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
