Assay Details
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139272.2 | NP_644678.2 | 6 - 7 | 735 | 109 | |
GenBank mRNA | AF348968.1 | - | 6 - 7 | 603 | 109 | |
AK145122.1 | - | 6 - 7 | 613 | 109 | ||
AK149609.1 | - | 6 - 7 | 661 | 110 | ||
AK151536.1 | - | 6 - 7 | 526 | 109 | ||
BC007172.1 | - | 5 - 6 | 470 | 109 | ||
BC053063.1 | - | 5 - 6 | 578 | 5702572 | 109 | |
BC059818.1 | - | 6 - 7 | 680 | 5697789 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
