Assay Details
Gene Symbol: Lpcat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145130.2 | NP_660112.1 | 6 - 7 | 784 | 64 | |
GenBank mRNA | AB294194.1 | - | 6 - 7 | 679 | 64 | |
AK083687.1 | - | 4 - 5 | 340 | 64 | ||
AK098090.1 | - | 6 - 7 | 755 | 64 | ||
AK154062.1 | - | 6 - 7 | 779 | 64 | ||
AK165319.1 | - | 6 - 7 | 763 | 64 | ||
AY028317.1 | - | 6 - 7 | 760 | 64 | ||
BC006753.1 | - | 6 - 7 | 712 | 3709076 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
